Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012404.2 | NP_036536.2 | 1 | 102 | ||
GenBank mRNA | BC069122.1 | - | 1 | 102 | 7216976 |