Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 9 | 1496 | ||
NM_001270492.1 | NP_001257421.1 | 10 | 1620 | |||
NM_001323527.1 | NP_001310456.1 | 10 | 1620 | |||
NM_001323529.1 | NP_001310458.1 | 9 | 1496 | |||
NM_001323533.1 | NP_001310462.1 | 9 | 1496 | |||
NM_001323534.1 | NP_001310463.1 | 10 | 1511 | |||
NM_001323535.1 | NP_001310464.1 | 9 | 1387 | |||
NM_139168.3 | NP_631907.1 | 10 | 1649 | |||
XM_011543171.2 | XP_011541473.1 | 9 | 1721 | |||
XM_017009056.1 | XP_016864545.1 | 9 | 1281 | |||
GenBank mRNA | AB209694.1 | - | 2 | 6790 | ||
AF459094.1 | - | 9 | 1330 | |||
AK091758.1 | - | 9 | 1379 | |||
AK125893.1 | - | 2 | 1181 | |||
AK308287.1 | - | 10 | 1523 | |||
BC067770.1 | - | 10 | 1650 | 4827589 | ||
BC112343.1 | - | 7 | 1018 | 40008558 |