Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300760.1 | NP_001287689.1 | 5 | 581 | ||
NM_139240.3 | NP_640333.3 | 7 | 680 | |||
XM_005245608.2 | XP_005245665.1 | 7 | 544 | |||
XM_005245610.3 | XP_005245667.1 | 6 | 461 | |||
XM_006711630.2 | XP_006711693.1 | 5 | 367 | |||
XM_011510152.2 | XP_011508454.1 | 9 | 2889 | |||
XM_011510153.2 | XP_011508455.1 | 8 | 2852 | |||
XM_011510154.2 | XP_011508456.1 | 6 | 2722 | |||
XM_011510155.2 | XP_011508457.1 | 8 | 2821 | |||
XM_011510156.2 | XP_011508458.1 | 5 | 2631 | |||
XM_017002808.1 | XP_016858297.1 | 7 | 478 | |||
GenBank mRNA | AL035295.1 | - | 7 | 680 | ||
BC038410.1 | - | 7 | 678 | 5169137 |