Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317911.1 | NP_001304840.1 | 3, 4 | 411 | ||
NM_001317912.1 | NP_001304841.1 | 4, 5 | 511 | |||
NM_139245.3 | NP_640338.2 | 3, 4 | 835 | |||
NR_134243.1 | 5, 6 | 750 | ||||
GenBank mRNA | AK055115.1 | - | 4, 5 | 511 | ||
AY337264.1 | - | 3, 4 | 831 | |||
BC104885.1 | - | 3, 4 | 397 | 8143888 | ||
BC104887.1 | - | 3, 4 | 397 | 8143890 |