Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321265.1 | NP_001308194.1 | 12 | 2042 | ||
NM_001321266.1 | NP_001308195.1 | 11 | 1964 | |||
NM_001321267.1 | NP_001308196.1 | 14 | 2265 | |||
NM_138799.3 | NP_620154.2 | 13 | 2188 | |||
NR_135598.1 | 14 | 2240 | ||||
NR_135599.1 | 13 | 2507 | ||||
NR_135600.1 | 13 | 2094 | ||||
NR_135601.1 | 11 | 1883 | ||||
NR_135602.1 | 12 | 2055 | ||||
NR_135603.1 | 14 | 2281 | ||||
NR_135604.1 | 13 | 2204 | ||||
NR_135605.1 | 12 | 2016 | ||||
NR_135606.1 | 13 | 2107 | ||||
GenBank mRNA | AK027321.2 | - | 13 | 2147 | ||
AK074779.1 | - | 3 | 1008 | |||
BC016005.1 | - | 10 | 1867 | |||
BC146871.1 | - | 13 | 2133 | 9056799 | ||
BC157827.1 | - | 13 | 2135 | 9057046 |