Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281293.1 | NP_001268222.1 | 2 | 283 | ||
NM_012482.4 | NP_036614.1 | 2 | 279 | |||
XM_017000887.1 | XP_016856376.1 | 5 | 1664 | |||
XM_017000888.1 | XP_016856377.1 | 5 | 622 | |||
GenBank mRNA | AF125158.1 | - | 1 | 175 | ||
AJ132592.1 | - | 2 | 259 | |||
AK022916.1 | - | 2 | 210 | |||
AK027284.1 | - | 2 | 277 | |||
AK297233.1 | - | 2 | 209 | |||
BC051905.1 | - | 1 | 168 | 6165292 | ||
BC060820.1 | - | 2 | 259 | 30344180 | ||
BP216938.1 | - | 2 | 283 |