Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138809.3 | NP_620164.1 | 6 | 1163 | ||
XM_017009034.1 | XP_016864523.1 | 6 | 927 | |||
XM_017009035.1 | XP_016864524.1 | 6 | 883 | |||
GenBank mRNA | AJ278125.1 | - | 6 | 956 | ||
AK074197.1 | - | 6 | 954 | |||
AK315390.1 | - | 5 | 717 | |||
BC001573.1 | - | 6 | 869 | 3454199 | ||
BX647141.1 | - | 4 | 435 |