Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178111.1 | NP_001171582.1 | 8 | 2113 | ||
NM_001178112.1 | NP_001171583.1 | 8 | 2080 | |||
NM_138967.3 | NP_620417.1 | 7 | 2030 | |||
NR_033660.1 | 6 | 1873 | ||||
XM_006720420.1 | XP_006720483.1 | 7 | 2189 | |||
XM_006720421.1 | XP_006720484.1 | 8 | 2071 | |||
GenBank mRNA | AB105192.1 | - | 7 | 2001 | ||
AF495715.1 | - | 7 | 1982 | |||
AK025678.1 | - | 1 | 840 | |||
AK056438.1 | - | 8 | 2045 | |||
AK126940.1 | - | 7 | 2030 | |||
AK127480.1 | - | 8 | 2050 | |||
AL360203.1 | - | 1 | 607 | |||
AL833230.1 | - | 1 | 1196 | |||
AL834226.1 | - | 7 | 2027 | |||
BC024700.1 | - | 6 | 1873 | 4941050 | ||
CR936761.1 | - | 7 | 1963 |