Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289082.1 | NP_001276011.1 | 7 | 1827 | ||
NM_001289083.1 | NP_001276012.1 | 8 | 1812 | |||
NM_001289084.1 | NP_001276013.1 | 7 | 1811 | |||
NM_001289085.1 | NP_001276014.1 | 8 | 1940 | |||
NM_001289086.1 | NP_001276015.1 | 7 | 1962 | |||
NM_001289087.1 | NP_001276016.1 | 6 | 1743 | |||
NM_139018.4 | NP_620587.2 | 7 | 1766 | |||
NR_110298.1 | 6 | 1804 | ||||
XM_017024212.1 | XP_016879701.1 | 7 | 1875 | |||
XM_017024213.1 | XP_016879702.1 | 8 | 1937 | |||
XM_017024214.1 | XP_016879703.1 | 8 | 1927 | |||
XM_017024215.1 | XP_016879704.1 | 8 | 1821 | |||
GenBank mRNA | AF251706.1 | - | 7 | 1638 | ||
AK092757.1 | - | 8 | 1766 | |||
AY358545.1 | - | 7 | 1713 | |||
BC028199.1 | - | 7 | 1805 | 5207520 |