Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139057.3 | NP_620688.2 | 6, 7 | 1145 | ||
XM_005254872.3 | XP_005254929.1 | 6, 7 | 2061 | |||
XM_011521312.2 | XP_011519614.1 | 6, 7 | 2061 | |||
XM_017021973.1 | XP_016877462.1 | 6, 7 | 2062 | |||
XM_017021974.1 | XP_016877463.1 | 6, 7 | 2064 | |||
XM_017021975.1 | XP_016877464.1 | 6, 7 | 2062 | |||
XM_017021976.1 | XP_016877465.1 | 5, 6 | 774 | |||
XM_017021977.1 | XP_016877466.1 | 6, 7 | 2061 | |||
XM_017021981.1 | XP_016877470.1 | 6, 7 | 2059 | |||
XM_017021984.1 | XP_016877473.1 | 5, 6 | 720 | |||
GenBank mRNA | AJ315735.1 | - | 6, 7 | 1062 | ||
AK131344.1 | - | 5, 6 | 691 |