Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 12 | 2397 | ||
NM_001270492.1 | NP_001257421.1 | 13 | 2518 | |||
NM_001323527.1 | NP_001310456.1 | 13 | 2521 | |||
NM_001323529.1 | NP_001310458.1 | 12 | 2394 | |||
NM_001323533.1 | NP_001310462.1 | 11 | 2252 | |||
NM_001323534.1 | NP_001310463.1 | 13 | 2412 | |||
NM_001323535.1 | NP_001310464.1 | 12 | 2285 | |||
NM_139168.3 | NP_631907.1 | 13 | 2550 | |||
XM_011543171.2 | XP_011541473.1 | 12 | 2622 | |||
XM_017009055.1 | XP_016864544.1 | 11 | 1931 | |||
XM_017009056.1 | XP_016864545.1 | 12 | 2182 | |||
GenBank mRNA | AF459094.1 | - | 12 | 2231 | ||
AK125893.1 | - | 5 | 2082 | |||
BC112343.1 | - | 10 | 1919 | 40008558 |