Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPTC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139283.1 | NP_644812.1 | 4 | 944 | ||
GenBank mRNA | AF385435.1 | - | 4 | 944 | ||
AK124744.1 | - | 5 | 788 | |||
AY357944.1 | - | 4 | 713 | |||
BC111551.1 | - | 5 | 746 | 40009728 | ||
CR749216.1 | - | 1 | 66 |