Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YME1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253866.1 | NP_001240795.1 | 8 | 983 | ||
NM_014263.3 | NP_055078.1 | 9 | 1082 | |||
NM_139312.2 | NP_647473.1 | 10 | 1253 | |||
XM_011519300.2 | XP_011517602.1 | 9 | 1536 | |||
GenBank mRNA | AF070656.1 | - | 5 | 397 | ||
AF151782.1 | - | 10 | 1097 | |||
AJ132637.1 | - | 9 | 1055 | |||
AK291292.1 | - | 9 | 1011 | |||
AK297973.1 | - | 8 | 947 | |||
AK309708.1 | - | 5 | 529 | |||
AK314372.1 | - | 9 | 1010 | |||
AY358484.1 | - | 9 | 1008 | |||
BC007795.2 | - | 10 | 1189 | 4110395 | ||
BC023507.2 | - | 9 | 1024 | 3637576 | ||
BC024032.2 | - | 9 | 1029 | 2961446 |