Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144582.2 | NP_653183.2 | 4, 5 | 494 | ||
GenBank mRNA | AK074840.1 | - | 2, 3 | 144 | ||
AK303594.1 | - | 4, 5 | 449 | |||
AK315543.1 | - | 4, 5 | 374 | |||
AY358783.1 | - | 4, 5 | 454 | |||
BC010656.1 | - | 2, 3 | 132 | |||
BC020251.1 | - | 2, 3 | 246 | |||
BC128461.1 | - | 4, 5 | 445 | 40126644 | ||
BC128462.1 | - | 4, 5 | 445 | 40126645 | ||
BI552105.1 | - | 4, 5 | 499 |