Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276326.1 | NP_001263255.1 | 5, 6 | 1607 | ||
NM_001276327.1 | NP_001263256.1 | 3, 4 | 1385 | |||
NM_144585.3 | NP_653186.2 | 5, 6 | 1709 | |||
NM_153378.2 | NP_700357.1 | 5, 6 | 1443 | |||
XM_006718430.3 | XP_006718493.1 | 4, 5 | 1390 | |||
XM_006718431.3 | XP_006718494.1 | 5, 6 | 1276 | |||
GenBank mRNA | AB050269.1 | - | 5, 6 | 1706 | ||
AB071863.1 | - | 5, 6 | 1109 | |||
AK055737.1 | - | 5, 6 | 1025 | |||
AK122599.1 | - | 5, 6 | 1263 | |||
AK309588.1 | - | 3, 4 | 320 | |||
AK315061.1 | - | 5, 6 | 1155 | |||
AY358183.1 | - | 3, 4 | 918 | |||
BC053348.1 | - | 5, 6 | 1252 | 5183650 | ||
DQ655911.2 | - | 91 | ||||
DQ655912.2 | - | 6, 7 | 600 |