Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144589.2 | NP_653190.2 | 7 | 931 | ||
GenBank mRNA | AK074421.1 | - | 7 | 884 | ||
AK290452.1 | - | 7 | 985 | |||
AK302283.1 | - | 7 | 841 | |||
AY358476.1 | - | 7 | 930 | |||
BC023663.2 | - | 7 | 852 | 4897624 | ||
BC047774.1 | - | 7 | 931 | 5761696 |