Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160113.1 | NP_001153585.1 | 3 | 294 | ||
NM_001160114.1 | NP_001153586.1 | 3 | 294 | |||
NM_001160115.1 | NP_001153587.1 | 3 | 294 | |||
NM_001160116.1 | NP_001153588.1 | 3 | 294 | |||
NM_144597.2 | NP_653198.2 | 3 | 294 | |||
NR_027649.1 | 3 | 305 | ||||
NR_027650.1 | 3 | 305 | ||||
XM_006720385.2 | XP_006720448.1 | 3 | 951 | |||
XM_011521212.2 | XP_011519514.1 | 3 | 951 | |||
XM_011521213.2 | XP_011519515.1 | 3 | 951 | |||
XM_011521214.2 | XP_011519516.1 | 3 | 951 | |||
GenBank mRNA | AJ705316.1 | - | 3 | 208 | ||
AK125586.1 | - | 2 | 1380 | |||
AK311484.1 | - | 3 | 256 | |||
AK312160.1 | - | 3 | 268 | |||
BC019820.1 | - | 3 | 259 | 4449390 | ||
BE906569.1 | - | 3 | 238 | |||
BQ185982.1 | - | 3 | 245 | |||
BY800475.1 | - | 3 | 227 | |||
CN347153.1 | - | 3 | 213 | |||
DC428269.1 | - | 3 | 292 | |||
DW416709.1 | - | 3 | 293 | |||
KU178855.1 | - | 3 | 179 |