Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144649.2 | NP_653250.2 | 4, 5 | 537 | ||
GenBank mRNA | AK057631.1 | - | 4, 5 | 451 | ||
BC062592.1 | - | 4, 5 | 432 | 5224669 |