Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278649.1 | NP_001265578.1 | 6 | 634 | ||
NM_014835.3 | NP_055650.1 | 6 | 653 | |||
NM_144498.2 | NP_653081.1 | 6 | 689 | |||
XM_017028163.1 | XP_016883652.1 | 6 | 689 | |||
XM_017028164.1 | XP_016883653.1 | 7 | 994 | |||
XM_017028165.1 | XP_016883654.1 | 7 | 994 | |||
XM_017028166.1 | XP_016883655.1 | 7 | 743 | |||
XM_017028167.1 | XP_016883656.1 | 7 | 757 | |||
XM_017028168.1 | XP_016883657.1 | 6 | 689 | |||
XM_017028169.1 | XP_016883658.1 | 6 | 613 | |||
XM_017028170.1 | XP_016883659.1 | 6 | 613 | |||
GenBank mRNA | AB018315.1 | - | 6 | 559 | ||
AF392447.1 | - | 6 | 650 | |||
AK000230.1 | - | 6 | 650 | |||
AK291851.1 | - | 6 | 617 | |||
AK296595.1 | - | 6 | 595 | |||
AK315576.1 | - | 6 | 650 | |||
AY028168.1 | - | 6 | 559 | |||
BC000296.2 | - | 6 | 593 | 2819652 | ||
BC004455.1 | - | 6 | 593 | 2819652 | ||
BC018812.2 | - | 6 | 593 | |||
CR456722.1 | - | 5 | 448 |