Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144563.2 | NP_653164.2 | 6, 7 | 645 | ||
GenBank mRNA | AY050633.1 | - | 6, 7 | 621 | ||
BC015529.1 | - | 6, 7 | 617 | 3904718 | ||
L35035.1 | - | 1, 2 | 49 |