Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM55B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100814.2 | NP_001094284.1 | 3 | 671 | ||
NM_144568.3 | NP_653169.2 | 3 | 650 | |||
GenBank mRNA | AK314021.1 | - | 3 | 651 | ||
AL137727.1 | - | 1 | 1133 | |||
BC002867.1 | - | 3 | 432 | 3940519 | ||
BC020947.1 | - | 3 | 515 | 4816227 | ||
BX161490.1 | - | 3 | 516 | |||
BX248025.1 | - | 3 | 480 |