Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281987.1 | NP_001268916.1 | 7 | 1935 | ||
NM_001281988.1 | NP_001268917.1 | 6 | 513 | |||
NM_144569.5 | NP_653170.3 | 7 | 1935 | |||
XM_011542413.2 | XP_011540715.1 | 7 | 504 | |||
XM_011542414.2 | XP_011540716.1 | 6 | 699 | |||
XM_017002779.1 | XP_016858268.1 | 6 | 1844 | |||
XM_017002780.1 | XP_016858269.1 | 5 | 1802 | |||
XM_017002781.1 | XP_016858270.1 | 6 | 1844 | |||
XM_017002782.1 | XP_016858271.1 | 6 | 1073 | |||
GenBank mRNA | AK091016.1 | - | 6 | 484 | ||
AK131447.1 | - | 7 | 1864 | |||
BC114430.1 | - | 7 | 1861 | 40038407 |