Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144570.2 | NP_653171.1 | 3 | 309 | ||
GenBank mRNA | AK023154.1 | - | 3 | 307 | ||
AK296888.1 | - | 3 | 273 | |||
AK296998.1 | - | 4 | 397 | |||
AK304025.1 | - | 3 | 391 | |||
AK309909.1 | - | 2 | 181 | |||
AK316514.1 | - | 3 | 365 | |||
AY323978.1 | - | 2 | 231 | |||
AY323979.1 | - | 3 | 319 | |||
AY598330.1 | - | 3 | 273 | |||
BC014438.1 | - | 3 | 284 | 4877757 | ||
BC060853.1 | - | 3 | 303 | 5296086 | ||
BM837125.1 | - | 3 | 330 |