Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144575.2 | NP_653176.2 | 5 | 628 | ||
XM_011533159.2 | XP_011531461.1 | 5 | 625 | |||
XM_011533160.2 | XP_011531462.1 | 6 | 816 | |||
XM_011533161.2 | XP_011531463.1 | 5 | 625 | |||
XM_011533163.2 | XP_011531465.1 | 5 | 625 | |||
XM_017005264.1 | XP_016860753.1 | 6 | 816 | |||
XM_017005265.1 | XP_016860754.1 | 5 | 625 | |||
XM_017005266.1 | XP_016860755.1 | 5 | 625 | |||
GenBank mRNA | AK074418.1 | - | 6 | 695 | ||
AK290298.1 | - | 5 | 629 | |||
AK308037.1 | - | 5 | 605 | |||
BC117345.1 | - | 5 | 589 | 40125896 | ||
BC117347.1 | - | 5 | 589 | 40125898 | ||
BX640793.1 | - | 5 | 628 |