Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144577.3 | NP_653178.3 | 9, 10 | 1648 | ||
XM_005259413.3 | XP_005259470.1 | 11, 12 | 1356 | |||
XM_005259414.3 | XP_005259471.1 | 11, 12 | 1352 | |||
XM_005259415.3 | XP_005259472.1 | 11, 12 | 1370 | |||
XM_005259416.4 | XP_005259473.1 | 4, 5 | 503 | |||
XM_011527515.2 | XP_011525817.1 | 12, 13 | 2299 | |||
XM_011527516.2 | XP_011525818.1 | 10, 11 | 1089 | |||
XM_011527517.1 | XP_011525819.1 | 11, 12 | 1352 | |||
XM_017027483.1 | XP_016882972.1 | 10, 11 | 2129 | |||
GenBank mRNA | AK057357.1 | - | 9, 10 | 1295 | ||
AK057488.1 | - | 3, 4 | 346 | |||
AK128144.1 | - | 11, 12 | 1622 | |||
AL122083.1 | - | 3, 4 | 267 | |||
BC007730.2 | - | 9, 10 | 1648 | |||
BC025752.1 | - | 9, 10 | 1093 | |||
BC117431.1 | - | 3, 4 | 446 | 40125982 | ||
BC117433.1 | - | 3, 4 | 446 | 40125984 |