Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HENMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102592.1 | NP_001096062.1 | 4 | 489 | ||
NM_144584.2 | NP_653185.2 | 4 | 672 | |||
XM_005270411.1 | XP_005270468.1 | 3 | 371 | |||
XM_017000179.1 | XP_016855668.1 | 3 | 473 | |||
GenBank mRNA | AK055087.1 | - | 4 | 672 | ||
AK310152.1 | - | 4 | 662 | |||
AK311542.1 | - | 3 | 473 | |||
BC012198.1 | - | 4 | 398 | 2906200 | ||
BC088366.1 | - | 4 | 409 | 30390911 | ||
BP378034.1 | - | 4 | 489 |