Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276326.1 | NP_001263255.1 | 9, 10 | 2252 | ||
NM_001276327.1 | NP_001263256.1 | 7, 8 | 2030 | |||
NM_144585.3 | NP_653186.2 | 9, 10 | 2354 | |||
NM_153378.2 | NP_700357.1 | 9, 10 | 2088 | |||
XM_006718430.3 | XP_006718493.1 | 8, 9 | 2035 | |||
XM_006718431.3 | XP_006718494.1 | 9, 10 | 1921 | |||
GenBank mRNA | AB050269.1 | - | 9, 10 | 2351 | ||
AB071863.1 | - | 9, 10 | 1754 | |||
AK122599.1 | - | 9, 10 | 1908 | |||
AY358183.1 | - | 7, 8 | 1563 | |||
BC053348.1 | - | 9, 10 | 1897 | 5183650 |