Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288781.1 | NP_001275710.1 | 5 | 566 | ||
NM_001288782.1 | NP_001275711.1 | 5 | 566 | |||
NM_001288783.1 | NP_001275712.1 | 5 | 566 | |||
NM_144596.3 | NP_653197.2 | 6 | 596 | |||
NM_198309.3 | NP_938051.1 | 5 | 566 | |||
NM_198310.3 | NP_938052.1 | 5 | 566 | |||
XM_006720035.1 | XP_006720098.1 | 4 | 848 | |||
XM_006720037.2 | XP_006720100.1 | 4 | 848 | |||
XM_011536432.1 | XP_011534734.1 | 4 | 848 | |||
XM_011536433.1 | XP_011534735.1 | 4 | 848 | |||
XM_011536434.1 | XP_011534736.1 | 4 | 848 | |||
XM_011536435.2 | XP_011534737.1 | 3 | 252 | |||
XM_017020977.1 | XP_016876466.1 | 4 | 848 | |||
XM_017020978.1 | XP_016876467.1 | 4 | 848 | |||
GenBank mRNA | AK093891.1 | - | 4 | 444 | ||
AK124675.1 | - | 5 | 446 | |||
AK296077.1 | - | 4 | 427 | |||
AK315772.1 | - | 4 | 451 | |||
AL833901.1 | - | 4 | 390 | |||
AY366523.1 | - | 4 | 422 | |||
AY366524.1 | - | 4 | 422 | |||
AY373972.1 | - | 4 | 422 | |||
BC026351.1 | - | 4 | 422 | 4799330 | ||
BC095433.1 | - | 4 | 404 | 30368216 | ||
BG330023.1 | - | 443 | ||||
BX161472.1 | - | 4 | 406 | |||
BX247959.1 | - | 6 | 720 | |||
BX248248.1 | - | 2 | 98 | |||
KU178849.1 | - | 4 | 370 | |||
KU178850.1 | - | 4 | 370 | |||
KU178851.1 | - | 4 | 370 | |||
KU178853.1 | - | 4 | 370 |