Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304499.1 | NP_001291428.1 | 5 | 501 | ||
NM_001304500.1 | NP_001291429.1 | 3 | 231 | |||
NM_144600.3 | NP_653201.1 | 4 | 429 | |||
NR_130755.1 | 3 | 344 | ||||
NR_130756.1 | 5 | 529 | ||||
NR_130757.1 | 2 | 146 | ||||
GenBank mRNA | AK055715.1 | - | 4 | 372 | ||
AL832498.1 | - | 4 | 392 | |||
AY507846.1 | - | 4 | 385 | |||
BC022321.1 | - | 4 | 380 | 4702249 | ||
BI460301.1 | - | 5 | 467 | |||
BI550657.1 | - | 3 | 320 | |||
BU661163.1 | - | 2 | 97 | |||
CB991926.1 | - | 5 | 518 | |||
DA178808.1 | - | 5 | 473 | |||
DB103294.1 | - | 3 | 175 | |||
HY004717.1 | - | 4 | 430 |