Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144622.2 | NP_653223.2 | 9 | 1523 | ||
XM_011509188.2 | XP_011507490.1 | 7 | 986 | |||
XM_011509189.2 | XP_011507491.1 | 4 | 1375 | |||
GenBank mRNA | AK057496.1 | - | 9 | 1523 | ||
AK097468.1 | - | 7 | 937 | |||
AK097993.1 | - | 4 | 1195 | |||
BC111794.1 | - | 9 | 1502 |