Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM131A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171093.1 | NP_001164564.1 | 6 | 1125 | ||
NM_144635.4 | NP_653236.3 | 6 | 1243 | |||
XM_005247113.2 | XP_005247170.1 | 7 | 1273 | |||
XM_005247114.3 | XP_005247171.1 | 5 | 1069 | |||
XM_011512413.2 | XP_011510715.1 | 5 | 1311 | |||
GenBank mRNA | AY358435.1 | - | 3 | 2371 | ||
BC026221.1 | - | 5 | 882 | 4399325 | ||
BC037299.1 | - | 6 | 1122 | 4825140 |