Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRGUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144648.1 | NP_653249.1 | 4, 5 | 615 | ||
XM_017011749.1 | XP_016867238.1 | 4, 5 | 2058 | |||
GenBank mRNA | AK057348.1 | - | 4, 5 | 615 | ||
BC104897.1 | - | 4, 5 | 668 | 8143900 | ||
BC104899.1 | - | 4, 5 | 668 | 8143902 |