Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145153.1 | NP_001138625.1 | 5 | 695 | ||
NM_144649.2 | NP_653250.2 | 5 | 638 | |||
GenBank mRNA | AK057631.1 | - | 5 | 552 | ||
BC062592.1 | - | 5 | 533 | 5224669 | ||
BC105783.1 | - | 5 | 613 | 4669222 |