Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144669.2 | NP_653270.1 | 2, 3 | 373 | ||
NR_133646.1 | 2, 3 | 373 | ||||
XM_005253553.4 | XP_005253610.2 | 2, 3 | 484 | |||
XM_011537956.2 | XP_011536258.2 | 2, 3 | 484 | |||
XM_017018855.1 | XP_016874344.1 | 2, 3 | 484 | |||
XM_017018859.1 | XP_016874348.1 | 2, 3 | 484 | |||
GenBank mRNA | AK056540.1 | - | 2, 3 | 271 | ||
AK295333.1 | - | 2, 3 | 201 | |||
BC043528.1 | - | 2, 3 | 373 | 5168267 |