Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278649.1 | NP_001265578.1 | 9 | 1047 | ||
NM_014835.3 | NP_055650.1 | 9 | 1066 | |||
NM_144498.2 | NP_653081.1 | 9 | 1102 | |||
XM_017028163.1 | XP_016883652.1 | 9 | 1102 | |||
XM_017028164.1 | XP_016883653.1 | 10 | 1407 | |||
XM_017028165.1 | XP_016883654.1 | 10 | 1407 | |||
XM_017028166.1 | XP_016883655.1 | 10 | 1156 | |||
XM_017028167.1 | XP_016883656.1 | 10 | 1170 | |||
XM_017028168.1 | XP_016883657.1 | 9 | 1102 | |||
XM_017028169.1 | XP_016883658.1 | 9 | 1036 | |||
XM_017028170.1 | XP_016883659.1 | 9 | 1036 | |||
GenBank mRNA | AB018315.1 | - | 9 | 972 | ||
AF392447.1 | - | 9 | 1063 | |||
AK000230.1 | - | 9 | 1063 | |||
AK093592.1 | - | 3 | 1829 | |||
AK124174.1 | - | 3 | 423 | |||
AK291851.1 | - | 9 | 1030 | |||
AK296595.1 | - | 9 | 1008 | |||
AY028168.1 | - | 9 | 972 | |||
BC000296.2 | - | 9 | 1006 | 2819652 | ||
BC004455.1 | - | 9 | 1006 | 2819652 | ||
BC018812.2 | - | 9 | 1006 | |||
CR456722.1 | - | 8 | 861 |