Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281987.1 | NP_001268916.1 | 16 | 3225 | ||
NM_001281988.1 | NP_001268917.1 | 15 | 1803 | |||
NM_144569.5 | NP_653170.3 | 16 | 3264 | |||
XM_011542413.2 | XP_011540715.1 | 16 | 1794 | |||
XM_011542414.2 | XP_011540716.1 | 15 | 1989 | |||
XM_017002779.1 | XP_016858268.1 | 15 | 3131 | |||
XM_017002780.1 | XP_016858269.1 | 14 | 3092 | |||
XM_017002781.1 | XP_016858270.1 | 14 | 3008 | |||
XM_017002782.1 | XP_016858271.1 | 15 | 2363 | |||
GenBank mRNA | AK058077.1 | - | 6 | 805 | ||
AK091016.1 | - | 15 | 1774 | |||
AK131447.1 | - | 16 | 3193 | |||
BC007910.2 | - | 8 | 987 | |||
BC029069.1 | - | 6 | 1246 | |||
BC114430.1 | - | 16 | 3151 | 40038407 |