Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144570.2 | NP_653171.1 | 5 | 2334 | ||
GenBank mRNA | AK023154.1 | - | 5 | 2332 | ||
AY323978.1 | - | 4 | 2256 | |||
AY323979.1 | - | 5 | 2344 | |||
BC060853.1 | - | 5 | 2328 | 5296086 |