Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK1IP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144578.3 | NP_653179.1 | 4 | 1698 | ||
XM_011537362.2 | XP_011535664.2 | 4 | 2542 | |||
GenBank mRNA | AK025580.1 | - | 4 | 1662 | ||
AL834285.1 | - | 4 | 1681 | |||
BC015621.2 | - | 4 | 1659 | 4843370 | ||
U90909.1 | - | 1 | 693 |