Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144579.2 | NP_653180.1 | 14 | 1622 | ||
XM_005264648.1 | XP_005264705.1 | 14 | 1505 | |||
XM_006712147.1 | XP_006712210.1 | 12 | 1326 | |||
XM_011533186.2 | XP_011531488.1 | 16 | 3294 | |||
XM_011533187.2 | XP_011531489.1 | 15 | 3167 | |||
XM_011533188.2 | XP_011531490.1 | 15 | 3149 | |||
XM_011533189.1 | XP_011531491.1 | 15 | 1652 | |||
XM_011533190.2 | XP_011531492.1 | 16 | 2199 | |||
XM_011533192.2 | XP_011531494.1 | 16 | 2433 | |||
XM_011533194.2 | XP_011531496.1 | 14 | 3050 | |||
XM_011533195.2 | XP_011531497.1 | 13 | 2968 | |||
XM_011533196.2 | XP_011531498.1 | 15 | 2378 | |||
XM_011533197.1 | XP_011531499.1 | 15 | 1629 | |||
XM_017005340.1 | XP_016860829.1 | 14 | 3022 | |||
XM_017005341.1 | XP_016860830.1 | 13 | 2905 | |||
XM_017005342.1 | XP_016860831.1 | 13 | 1410 | |||
XM_017005343.1 | XP_016860832.1 | 13 | 2107 | |||
XM_017005344.1 | XP_016860833.1 | 14 | 2254 | |||
XM_017005345.1 | XP_016860834.1 | 7 | 2751 | |||
XM_017005346.1 | XP_016860835.1 | 15 | 1596 | |||
GenBank mRNA | AK094205.1 | - | 1 | 268 | ||
AK289731.1 | - | 14 | 1511 | |||
AK315861.1 | - | 3 | 730 | |||
AY044437.1 | - | 14 | 1626 | |||
BC013233.1 | - | 8 | 1061 |