Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190415.1 | NP_001177344.1 | 4 | 555 | ||
NM_005641.3 | NP_005632.1 | 4 | 884 | |||
NM_139315.2 | NP_647476.1 | 4 | 586 | |||
NR_033792.1 | 4 | 435 | ||||
XM_006716100.2 | XP_006716163.1 | 5 | 677 | |||
XM_006716101.2 | XP_006716164.1 | 4 | 514 | |||
XM_006716102.2 | XP_006716165.1 | 4 | 381 | |||
XM_011516542.2 | XP_011514844.1 | 5 | 656 | |||
XM_011516543.2 | XP_011514845.1 | 4 | 409 | |||
XM_017012568.1 | XP_016868057.1 | 4 | 854 | |||
GenBank mRNA | AK097781.1 | - | 5 | 494 | ||
AK300005.1 | - | 4 | 538 | |||
AK314428.1 | - | 4 | 861 | |||
AK315077.1 | - | 4 | 536 | |||
BC018115.1 | - | 4 | 529 | 3905205 | ||
CR627390.1 | - | 4 | 413 | |||
DA150478.1 | - | 4 | 514 | |||
L25444.1 | - | 4 | 856 | |||
U31659.1 | - | 3 | 640 |