Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HENMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102592.1 | NP_001096062.1 | 8 | 1035 | ||
NM_144584.2 | NP_653185.2 | 8 | 1218 | |||
XM_005270411.1 | XP_005270468.1 | 7 | 917 | |||
XM_017000179.1 | XP_016855668.1 | 7 | 1019 | |||
GenBank mRNA | AK055087.1 | - | 8 | 1218 | ||
BC012198.1 | - | 8 | 944 | 2906200 | ||
BC088366.1 | - | 8 | 955 | 30390911 |