Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144616.3 | NP_653217.1 | 7 | 838 | ||
XM_017026285.1 | XP_016881774.1 | 7 | 1253 | |||
XM_017026286.1 | XP_016881775.1 | 7 | 1232 | |||
XM_017026287.1 | XP_016881776.1 | 7 | 835 | |||
GenBank mRNA | AK056978.1 | - | 7 | 832 | ||
BC021201.2 | - | 6 | 802 | 4106867 |