Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039500.2 | NP_001034589.2 | 20 | 3201 | ||
XM_006710360.3 | XP_006710423.2 | 17 | 2476 | |||
XM_011540683.2 | XP_011538985.1 | 20 | 3163 | |||
XM_011540684.2 | XP_011538986.1 | 20 | 3288 | |||
XM_011540685.2 | XP_011538987.1 | 20 | 3300 | |||
XM_011540688.2 | XP_011538990.1 | 19 | 3088 | |||
XM_011540690.2 | XP_011538992.1 | 19 | 2986 | |||
XM_011540691.2 | XP_011538993.1 | 19 | 2977 | |||
XM_011540692.2 | XP_011538994.1 | 20 | 3157 | |||
XM_011540697.2 | XP_011538999.1 | 14 | 2534 | |||
XM_017000305.1 | XP_016855794.1 | 14 | 2280 | |||
XM_017000306.1 | XP_016855795.1 | 15 | 2441 | |||
GenBank mRNA | AK057346.1 | - | 10 | 1485 | ||
AK295797.1 | - | 20 | 3185 | |||
BC101378.1 | - | 9 | 1281 | |||
BC101379.2 | - | 8 | 1206 | |||
BC101380.1 | - | 9 | 1281 | |||
BC101381.1 | - | 9 | 1281 |