Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144625.4 | NP_653226.4 | 24 | 3146 | ||
XM_006711736.3 | XP_006711799.1 | 25 | 3176 | |||
XM_011544085.2 | XP_011542387.1 | 22 | 2575 | |||
XM_011544086.2 | XP_011542388.1 | 23 | 2577 | |||
XM_017000315.1 | XP_016855804.1 | 25 | 2911 | |||
GenBank mRNA | AK057540.1 | - | 12 | 1277 | ||
AK122927.1 | - | 24 | 2986 | |||
AY327404.1 | - | 14 | 1751 |