Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042625.1 | NP_001036090.1 | 4 | 568 | ||
NM_144647.3 | NP_653248.3 | 4 | 600 | |||
XM_006714444.3 | XP_006714507.1 | 4 | 550 | |||
XM_006714445.3 | XP_006714508.1 | 4 | 558 | |||
GenBank mRNA | BC017586.2 | - | 4 | 602 | 4837506 |