Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144658.3 | NP_653259.3 | 52 | 6114 | ||
XM_005262368.3 | XP_005262425.1 | 53 | 6398 | |||
XM_005262369.4 | XP_005262426.1 | 53 | 6092 | |||
XM_011531276.2 | XP_011529578.1 | 53 | 6033 | |||
XM_011531278.2 | XP_011529580.1 | 40 | 4519 | |||
GenBank mRNA | AK054649.1 | - | 13 | 1796 | ||
AK056684.1 | - | 22 | 2628 | |||
AK124735.1 | - | 47 | 5537 | |||
AK125641.1 | - | 31 | 3626 | |||
AY692226.1 | - | 52 | 6114 | |||
BC047713.1 | - | 18 | 2205 |