Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144669.2 | NP_653270.1 | 8 | 1414 | ||
NR_133646.1 | 11 | 1602 | ||||
XM_011537956.2 | XP_011536258.2 | 13 | 1813 | |||
XM_011537957.2 | XP_011536259.1 | 12 | 1502 | |||
XM_017018857.1 | XP_016874346.1 | 10 | 1378 | |||
XM_017018858.1 | XP_016874347.1 | 9 | 1321 | |||
XM_017018860.1 | XP_016874349.1 | 9 | 1282 | |||
XM_017018861.1 | XP_016874350.1 | 11 | 1447 | |||
GenBank mRNA | AK056540.1 | - | 8 | 1312 | ||
AK295333.1 | - | 13 | 1530 | |||
AK295454.1 | - | 10 | 1265 | |||
BC043528.1 | - | 11 | 1602 | 5168267 |