Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528605.2 | XP_011526907.1 | 5 | 909 | ||
XM_011528606.2 | XP_011526908.1 | 5 | 827 | |||
XM_011528607.2 | XP_011526909.1 | 5 | 913 | |||
XM_011528612.2 | XP_011526914.1 | 5 | 579 | |||
GenBank mRNA | AK058202.1 | - | 5 | 584 | ||
BC057387.1 | - | 4 | 2053 | 5724236 |