Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018060.2 | NP_001018070.1 | 4 | 502 | ||
NM_001146288.1 | NP_001139760.1 | 4 | 587 | |||
NM_144704.2 | NP_653305.1 | 4 | 502 | |||
GenBank mRNA | AK055035.1 | - | 4 | 486 | ||
AK094844.1 | - | 4 | 491 | |||
BC032485.1 | - | 4 | 469 | 5221203 | ||
CR456342.1 | - | 3 | 321 | |||
DA396585.1 | - | 4 | 524 |