Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144718.3 | NP_653319.1 | 4 | 532 | ||
XM_011512461.2 | XP_011510763.1 | 4 | 289 | |||
XM_017005775.1 | XP_016861264.1 | 4 | 289 | |||
XM_017005776.1 | XP_016861265.1 | 4 | 426 | |||
XM_017005777.1 | XP_016861266.1 | 4 | 349 | |||
XM_017005778.1 | XP_016861267.1 | 4 | 533 | |||
GenBank mRNA | AK057966.1 | - | 4 | 313 | ||
AK296160.1 | - | 4 | 426 | |||
AL832148.1 | - | 4 | 501 | |||
AL832327.2 | - | 4 | 315 | |||
AY099107.1 | - | 4 | 452 | |||
BC036951.1 | - | 4 | 473 | 5500345 | ||
BP278430.1 | - | 4 | 532 |